Learn More
Aviva Systems Biology Corp RPS6KB1 Antibody - N-terminal region (ARP56554_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP56554_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against RPS6KB1. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human RPS6KB1. Alternate names: PS6K, S6K, S6K1, STK14A, p70(S6K)-alpha, p70-S6K, p70-alphaApplicationWB. Species reactivity: Cow, Dog, Goat, Guinea Pig, Horse, Human, Mouse, Pig, Rabbit, Rat, Zebrafish
Catalog No. 50-014-61903