Learn More
Aviva Systems Biology Corp Rpsa Antibody - N-terminal region (ARP54682_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP54682_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against Rpsa. It was validated on Western Blot. The immunogen is a synthetic peptide corresponding to a region of Mouse. Alternate names: 67kDa, 67lr, AL022858, Lamr, Lamr1, Lamrl1, MGC117921, MGC118614, MLR, P40, P40-3, P40-8ApplicationWB. Species reactivity: Cow, Dog, Goat, Horse, Human, Mouse, Pig, Rabbit, Rat, Sheep, Zebrafish
Catalog No. 50-285-952