Learn More
USA Scientific Inc TipOne RPT Filter Tips
Supplier:  USA Scientific Inc 11808810
Encompass
TipOne RPT Filter Tips are sterile, single-use pipette tips with a built-in filter designed for preventing cross-contamination when pipetting. They feature a hydrophobic PTFE membrane that allows air to pass through while blocking aerosols and liquids. The filter is integrated into the tip near the end, with a 10μL dead volume. These filter tips come in a resealable bag, individually wrapped in medical-grade paper. They are made from medical grade virgin polypropylene and are guaranteed sterile and free of DNase, RNase and pyrogens. They are available in sizes from 10μL to 1000μL.
Catalog No. NC2177914