Learn More
Sigma Aldrich Fine Chemicals Biosciences RPTEC Complete Supplement
Catalog No. 502901611
Encompass
Sigma has developed a variety of specialized media which are intended to provide optimal performance when utilized with specific cell lines. This product consists of 30 mL of RPTEC complete media supplement which is added to basal media and other components listed below to produce 500 mL of complete media needed for optimal growth maintenance and culture of Sigmas Human Renal Proximal Tubule Epithelial Cells (RPTEC). The final RPTEC complete media was developed specifically for use with Sigmas human RPTEC control SA7K Clone cells and various knockouts (KO).
Catalog No. 50-290-1611 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. MTOXRCSUP30ML
Sigma has developed a variety of specialized media which are intended to provide optimal performance when utilized with specific cell lines. This product consists of 30 mL of RPTEC complete media supplement which is added to basal media and other components listed below to produce 500 mL of complete media needed for optimal growth maintenance and culture of Sigmas Human Renal Proximal Tubule Epithelial Cells (RPTEC). The final RPTEC complete media was developed specifically for use with Sigmas human RPTEC control SA7K Clone cells and various knockouts (KO).