Learn More
Lonza Walkersville RPTEC - Human Renal Proximal Tubule Epithelial Cells
Catalog No. NC9760462
Encompass
Lonza's Human Renal Cells include cells isolated from various normal human kidney tissue specific to each cell type.Human Renal Proximal Tubule Cells are isolated from the proximal tubule and are ideal for drug transporter studies. Human Renal Epithelial Cells (HRE) are comprised of cells from the cortex and glomerular region. Human Renal Cortical Cells (HRCE) are from proximal and distal tubules. Human Mesangial Cells (HMC) are isolated from the renal glomerulus and modified smooth muscle cells between capillaries.Benefits:Guaranteed through 15 population doublings Cells stain positive for gamma GTP and pan cytokeratinTest negative for mycoplasma, bacteria, yeast, and fungiHIV-1, Hepatitis B and Hepatitis C are not detected for all donors and/or cell lotsA Certificate ofAnalysis is provided for each cell lot purchasedContent:1 x Cryopreserved ampule of RPTEC containing ≥500,000 cells
Catalog No. NC9760462 Supplier Lonza Walkersville Supplier No. CC2553
Lonza's Human Renal Cells include cells isolated from various normal human kidney tissue specific to each cell type.Human Renal Proximal Tubule Cells are isolated from the proximal tubule and are ideal for drug transporter studies. Human Renal Epithelial Cells (HRE) are comprised of cells from the cortex and glomerular region. Human Renal Cortical Cells (HRCE) are from proximal and distal tubules. Human Mesangial Cells (HMC) are isolated from the renal glomerulus and modified smooth muscle cells between capillaries.Benefits:Guaranteed through 15 population doublings Cells stain positive for gamma GTP and pan cytokeratinTest negative for mycoplasma, bacteria, yeast, and fungiHIV-1, Hepatitis B and Hepatitis C are not detected for all donors and/or cell lotsA Certificate ofAnalysis is provided for each cell lot purchasedContent:1 x Cryopreserved ampule of RPTEC containing ≥500,000 cells