Learn More
Eurofins Discoverx RPTPµ, active, 250 µg. (14-780M)
Supplier:  Eurofins Discoverx 14780M
Encompass
The RPTPµ, active product is purified N terminal GST-tagged recombinant RPTPµ enzyme. This enzyme contains amino acids 879-1184. It retains its binding and enzymatic activity and is intended for use in phosphatase assays. This enzyme is provided as 10 μg of 0.93 mg/mL frozen solution. For detailed product information, visit www.discoverx.com/proteins.
Catalog No. 50-199-9332