Learn More
Cayman Chemical RpUrIdIn5O 1thIotrIpho 2.5mg
SDP
Catalog No. 502918829
Encompass_Preferred
A nucleotide P2Y2 and P2Y4 receptor agonist; induces inositol phosphate accumulation in 1321N1 cells expressing P2Y2 or P2Y4 receptors (EC50s = 5.4 and 27 µM, respectively)
Catalog No. 50-291-8829 Supplier Cayman Chemical Supplier No. 385722.5 MG
Add to Cart
Add to Cart
A nucleotide P2Y2 and P2Y4 receptor agonist; induces inositol phosphate accumulation in 1321N1 cells expressing P2Y2 or P2Y4 receptors (EC50s = 5.4 and 27 µM, respectively)