Learn More
Aviva Systems Biology Corp RPUSD3 Antibody - middle region (ARP41173_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP41173_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against RPUSD3. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human RPUSD3. Alternate names: FLJ34707, MGC29784ApplicationWB. Species reactivity: Cow, Dog, Human, Mouse, Pig, Rat
Catalog No. 50-014-57749