Learn More
Cayman Chemical RQ-00203078, Purity: ≥98%, CAS: 1254205-52-1, 5 mg
SDP
Catalog No. 502982350
Encompass_Preferred
A selective TRPM8 antagonist (IC50s = 8.3 and 5.8 nM in human and rat, respectively); attenuates icilin-induced wet-dog shakes in rats (ED50 = 0.65 mg/kg); reduces HSC3 and HSC4 oral squamous carcinoma cell migration and invasion in vitro
Catalog No. 50-298-2350 Supplier Cayman Chemical Supplier No. 190755 MG
A selective TRPM8 antagonist (IC50s = 8.3 and 5.8 nM in human and rat, respectively); attenuates icilin-induced wet-dog shakes in rats (ED50 = 0.65 mg/kg); reduces HSC3 and HSC4 oral squamous carcinoma cell migration and invasion in vitro