Learn More
Medchemexpress LLC HY-112205A 10mg Medchemexpress, RR-11a analog CAS:685543-66-2 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY112205A10MG
Encompass_Preferred
Medchemexpress, HY-112205A 10mg RR-11a analog CAS:685543-66-2 RR-11a analog is a potent and selective inhibitors of asparaginyl endopeptidases (AE) (Legumain), with IC50 values of 4.5 nM, 4.5 nM and 31 nM for AE1 in Trichomonas Vaginalis, AE in Ixodes ricinus and AE in Schistosoma mansoni, respectively. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-196-9059