Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC RR-11a analog | 685543-66-2 | 99.1% | 491.49 | 1 ML
SDP

Catalog No. 5000438337
Encompass_Preferred

RR-11a analog is a potent and irreversible inhibitor of *Schistosoma mansoni* legumain, with an IC50 of 31 nM. It is an aza-Asn derivative and an aza-peptide Michael acceptor. For research use only.

  • Potent and irreversible inhibitor of *Schistosoma mansoni* legumain
  • Aza-Asn derivative and aza-peptide Michael acceptor
  • Molecular weight: 491.49
  • Formula: C22H29N5O8
  • Appearance: Solid
  • Color: White to off-white
  • Solubility in DMSO: 125 mg/mL (254.33 mM; requires ultrasonic)
  • Solubility in H2O: 2.5 mg/mL (5.09 mM; requires ultrasonic and warming to 60°C)

Catalog No. 50-004-38337 Supplier Medchemexpress LLC Supplier No. HY112205A10MM/1ML
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

RR-11a analog is a potent and irreversible inhibitor of *Schistosoma mansoni* legumain, with an IC50 of 31 nM. It is an aza-Asn derivative and an aza-peptide Michael acceptor. For research use only.

  • Potent and irreversible inhibitor of *Schistosoma mansoni* legumain
  • Aza-Asn derivative and aza-peptide Michael acceptor
  • Molecular weight: 491.49
  • Formula: C22H29N5O8
  • Appearance: Solid
  • Color: White to off-white
  • Solubility in DMSO: 125 mg/mL (254.33 mM; requires ultrasonic)
  • Solubility in H2O: 2.5 mg/mL (5.09 mM; requires ultrasonic and warming to 60°C)

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.