Learn More
Strem, An Ascensus Company CAS 71042-55-2. 250mg. (2R3R)-(-)-23-Bis(diphenylphosphino)-bicyclo[2.2.1]hept-5-ene min. 95% (RR)-NORPHOS. MFCD00085365
SDP
Catalog No. 502110727
Encompass
CAS 71042-55-2. 250mg. (2R3R)-(-)-23-Bis(diphenylphosphino)-bicyclo[2.2.1]hept-5-ene min. 95% (RR)-NORPHOS. MFCD00085365. Molecular Weight 462.51. Molecular Formula C31H28P2. Color/form white xtl. Strem 15-0140. http//www.strem.com/catalog/v/15-0140/
Catalog No. 50-211-0727 Supplier Strem, An Ascensus Company Supplier No. 150140250MG
CAS 71042-55-2. 250mg. (2R3R)-(-)-23-Bis(diphenylphosphino)-bicyclo[2.2.1]hept-5-ene min. 95% (RR)-NORPHOS. MFCD00085365. Molecular Weight 462.51. Molecular Formula C31H28P2. Color/form white xtl. Strem 15-0140. http//www.strem.com/catalog/v/15-0140/