Learn More
Strem, An Ascensus Company CAS 521272-85-5. 100mg. (2R3R)-(-)-23-Bis(diphenylphosphino)bicyclo[2.2.1]hept-5-ene(15-cyclooctadiene)rhodium(I) tetrafluoroborate min. 97% (RR)-NORPHOS-Rh. MFCD09839137
SDP
Catalog No. 502110838
Encompass
CAS 521272-85-5. 100mg. (2R3R)-(-)-23-Bis(diphenylphosphino)bicyclo[2.2.1]hept-5-ene(15-cyclooctadiene)rhodium(I) tetrafluoroborate min. 97% (RR)-NORPHOS-Rh. MFCD09839137. Molecular Weight 760.39. Molecular Formula [Rh(C8H12)(C31H28P2)]BF4ae. Color/form red-orange pwdr. Strem 45-0184. http//www.strem.com/catalog/v/45-0184/
Catalog No. 50-211-0838 Supplier Strem, An Ascensus Company Supplier No. 450184100MG
CAS 521272-85-5. 100mg. (2R3R)-(-)-23-Bis(diphenylphosphino)bicyclo[2.2.1]hept-5-ene(15-cyclooctadiene)rhodium(I) tetrafluoroborate min. 97% (RR)-NORPHOS-Rh. MFCD09839137. Molecular Weight 760.39. Molecular Formula [Rh(C8H12)(C31H28P2)]BF4ae. Color/form red-orange pwdr. Strem 45-0184. http//www.strem.com/catalog/v/45-0184/