Learn More
Gold Biotechnology Inc RR1 Competent E coli Cells
SDP
Catalog No. 5000562197
Encompass_Preferred
GoldBio s RR1 chemically competent E coli cells are suitable for high-efficiency transformation in a wide variety of applications such as cloning and sub-cloning E coli RR1 is a recA derivative of the HB101 strain and can be more useful than HB101 if a recA background is required
Catalog No. 50-005-62197 Supplier Gold Biotechnology Inc Supplier No. CC11310X50
GoldBio s RR1 chemically competent E coli cells are suitable for high-efficiency transformation in a wide variety of applications such as cloning and sub-cloning E coli RR1 is a recA derivative of the HB101 strain and can be more useful than HB101 if a recA background is required