Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

RR688 Mouse Anti-Human IgE, BD OptiBuild™

Catalog No. BDB772766 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB772766 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB772766 Supplier BD Biosciences Supplier No. 772766
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Mouse Monoclonal Secondary Antibody

Monoclonal Antibody Specifically detects in Human Samples. It is validated for Flow Cytometry.

The G7-26 monoclonal antibody specifically recognizes the constant region of human Immunoglobulin E (IgE). It does not crossreact with other immunoglobulin heavy chain isotypes. IgE exists as a transmembrane form on B lymphocytes and as a soluble form produced by activated B cells and plasma cells. IgE can bind to cell surface receptors like high-affinity (FcεRI) on mast cells, basophils, dendritic cells, and monocytes, or low-affinity (FcεRII/CD23) on B cells, T cells, monocytes, eosinophils, neutrophils, follicular dendritic cells, and Langerhans cells. Crosslinking of Fc receptor-bound IgE by multivalent antigens or allergens can induce phagocytosis or release of inflammatory mediators. IgE provides immune protection against pathogenic parasites and plays a role in allergic disorders.
TRUSTED_SUSTAINABILITY

Specifications

Applications Flow Cytometry
Classification Monoclonal
Clone G7-26
Concentration 0.2 mg/mL
Conjugate RR688
Formulation Aqueous buffered solution containing ≤0.09% sodium azide.
Gene Alias Immunoglobulin E; IGHE; IGHε; Ig epsilon chain constant region
Host Species Mouse
Quantity 50 μg
Regulatory Status RUO
Primary or Secondary Secondary
Gene ID (Entrez) 3497
Target Species Human
Content And Storage Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. The monoclonal antibody was purified from tissue culture supernatant or ascites by affinity chromatography. The antibody was conjugated to the dye under optimum conditions that minimize unconjugated dye and antibody.
Product Type Antibody
Form Liquid
Isotype IgG2a κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.