Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

RR688 Mouse Anti-Human IgG1, BD OptiBuild™

Catalog No. BDB773002 Shop All Waters Corporation Products
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB773002 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB773002 Supplier BD Biosciences Supplier No. 773002
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Mouse Monoclonal Secondary Antibody

Monoclonal Antibody Specifically detects in Human Samples. It is validated for Flow Cytometry.

The HP6001 monoclonal antibody specifically recognizes the IgG1 subclass of human Immunoglobulin G (IgG). Human IgG1 is the most abundant IgG subclass in human serum. IgG1 consists of two identical heavy chains (encoded by IGHG1) and two light chains (Igκ or Igλ). The HP6001 antibody binds to the CH2 domain of the IgG1 heavy chain and does not cross-react with other immunoglobulin heavy chain (IgH) subclasses. IgG1 is expressed by plasmablasts, plasma cells, memory B cells, and some myeloma/plasmacytoma cells. HP6001 binds to soluble human IgG1, cytophilic IgG1 attached to cells via its Fc region, or human IgG1 antibodies specifically bound to antigens. IgG1 can cross the placenta and disseminate in extravascular fluids. Functions of human IgG1 include antibody-dependent neutralization of toxins or microbial infection, opsonization for phagocytosis, complement fixation, and cell-mediated cytotoxicity.
TRUSTED_SUSTAINABILITY

Specifications

Applications Flow Cytometry
Classification Monoclonal
Clone HP6001 (also known as FB5-EEI2)
Concentration 0.2 mg/mL
Conjugate RR688
Formulation Aqueous buffered solution containing ≤0.09% sodium azide.
Gene Alias IGHG1; IgG1 (Fc); Immunoglobulin gamma 1; Immunoglobulin heavy constant gamma 1
Host Species Mouse
Immunogen Purified Human Immunoglobulins
Quantity 50 μg
Regulatory Status RUO
Primary or Secondary Secondary
Gene ID (Entrez) 3500
Target Species Human
Content And Storage Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. The monoclonal antibody was purified from tissue culture supernatant or ascites by affinity chromatography. The antibody was conjugated to the dye under optimum conditions that minimize unconjugated dye and antibody.
Product Type Antibody
Form Liquid
Isotype IgG2b κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.