Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

RR688 Mouse Anti-Human IgG2, BD OptiBuild™

Catalog No. BDB772420 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB772420 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB772420 Supplier BD Biosciences Supplier No. 772420
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Mouse Monoclonal Secondary Antibody

Monoclonal Antibody Specifically detects in Human Samples. It is validated for Flow Cytometry.

The HP6002 monoclonal antibody specifically recognizes the subclass of human Immunoglobulin G (IgG) known as IgG2. It binds to the CH2 domain of the IgG2 heavy chain and does not cross-react with other immunoglobulin heavy chain (IgH) subclasses. IgG2 is expressed by plasmablasts, plasma cells, and memory B cells. HP6002 binds to soluble human IgG2, cytophilic IgG2 attached to cells through its Fc region, or human IgG2 antibodies specifically bound to antigens. Human IgG2 serves multiple functions, including antibody-dependent neutralization and complement fixation. The BD Horizon RealRed 688 (RR688) Dye is a small organic fluorochrome with an excitation maximum at 669 nm and an emission maximum at 688 nm, designed for detection on spectral cytometers and to be excited by the Red laser (627-640 nm).
TRUSTED_SUSTAINABILITY

Specifications

Applications Flow Cytometry
Classification Monoclonal
Clone HP6002 (also known as AB4-DH4)
Concentration 0.2 mg/mL
Conjugate RR688
Formulation Aqueous buffered solution containing ≤0.09% sodium azide.
Gene Alias IGHG2; Immunoglobulin heavy constant gamma 2
Host Species Mouse
Immunogen Purified Human IgG2 Myeloma Protein
Quantity 50 μg
Regulatory Status RUO
Primary or Secondary Secondary
Gene ID (Entrez) 3501
Target Species Human
Content And Storage Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. The monoclonal antibody was purified from tissue culture supernatant or ascites by affinity chromatography. The antibody was conjugated to the dye under optimum conditions that minimize unconjugated dye and antibody.
Product Type Antibody
Form Liquid
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.