Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

RR688 Mouse Anti-Human IgM, BD OptiBuild™

Catalog No. BDB773415 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB773415 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB773415 Supplier BD Biosciences Supplier No. 773415
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Mouse Monoclonal Secondary Antibody

Monoclonal Antibody Specifically detects in Human Samples. It is validated for Flow Cytometry.

The UCH-B1 monoclonal antibody specifically recognizes the heavy chain constant region (Cμ) of human Immunoglobulin M (IgM) encoded by IGHM. It does not crossreact with other immunoglobulin heavy or light chain isotypes. Intracytoplasmic IgM is expressed by pre-B cells, while immature and some mature B cells express cell surface IgM. Cell surface IgM consists of two type I transmembrane Igμ heavy chains paired with either Igκ or Igλ light chains, serving as a receptor for antigens. The UCH-B1 monoclonal antibody recognizes both cell surface and soluble human IgM. IgM is important in defense against pathogens and may play a role in autoimmune diseases. The UCH-B1 antibody can reportedly activate or costimulate the proliferation of normal B cells or some transformed B cell lines.
TRUSTED_SUSTAINABILITY

Specifications

Applications Flow Cytometry
Classification Monoclonal
Clone UCH-B1 (also known as UCHB1)
Concentration 0.2 mg/mL
Conjugate RR688
Formulation Aqueous buffered solution containing ≤0.09% sodium azide.
Gene Alias UCHB1; IGHM; MU
Host Species Mouse
Immunogen Human Prolymphocytic Leukemia B Cells
Quantity 50 μg
Regulatory Status RUO
Primary or Secondary Secondary
Gene ID (Entrez) 3507
Target Species Human
Content And Storage Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. The monoclonal antibody was purified from tissue culture supernatant or ascites by affinity chromatography. The antibody was conjugated to the dye under optimum conditions that minimize unconjugated dye and antibody.
Product Type Antibody
Form Liquid
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.