Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

RR688 Mouse Anti-Mouse IgD[a], BD OptiBuild™

Catalog No. BDB772674 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB772674 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB772674 Supplier BD Biosciences Supplier No. 772674
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Mouse Monoclonal Secondary Antibody

Monoclonal Antibody Specifically detects in Mouse Samples. It is validated for Flow Cytometry.

The AMS 9.1 antibody reacts specifically with the Fc portion of mouse IgD of Igh C[a] haplotype (e.g., AKR, BALB/c, CBA, C3H/He, C58, DBA/1, DBA/2, NZB) and does not react with IgD of Igh-C[b] or Igh-C[e] haplotypes (e.g., C57BL/6, C57BL/10, SJL, A). This mAb is effective for detection of cell-surface Ig by immunofluorescent staining with flow cytometric analysis and has been reported to stimulate B-cell proliferation. The BD Horizon RealRed 688 (RR688) Dye is a small organic fluorochrome with an excitation maximum at 669 nm and an emission maximum at 688 nm. RR688 is designed for detection on spectral cytometers and to be excited by the Red laser (627-640 nm). RR688 can be used on a spectral instrument in combination with APC and either R718 or Alexa Fluor 700.
TRUSTED_SUSTAINABILITY

Specifications

Applications Flow Cytometry
Classification Monoclonal
Clone AMS 9.1
Concentration 0.2 mg/mL
Conjugate RR688
Formulation Aqueous buffered solution containing ≤0.09% sodium azide.
Gene Alias Igh-5a; Igh-5.4; Ighd; Immunoglobulin D
Host Species Mouse
Immunogen BALB/c mouse splenocytes
Quantity 50 μg
Regulatory Status RUO
Primary or Secondary Secondary
Gene ID (Entrez) 380797
Target Species Mouse
Content And Storage Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. The monoclonal antibody was purified from tissue culture supernatant or ascites by affinity chromatography. The antibody was conjugated to the dye under optimum conditions that minimize unconjugated dye and antibody.
Product Type Antibody
Form Liquid
Isotype IgG2b κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.