Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

RR688 Mouse Anti-Rat Ig, λ Light Chain, BD OptiBuild™

Catalog No. BDB772775 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB772775 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB772775 Supplier BD Biosciences Supplier No. 772775
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Mouse Monoclonal Secondary Antibody

Monoclonal Antibody Specifically detects in Rat Samples. It is validated for Flow Cytometry.

The B46-5 monoclonal antibody specifically recognizes rat Igs bearing a λ light chain. It does not react with rat Ig κ light chain or heavy chains, nor with rabbit, hamster, guinea pig, or human immunoglobulins. In some cases, it may cross-react with rat IgE κ isotype antibodies. The BD Horizon RealRed 688 (RR688) Dye is a small organic fluorochrome with an excitation maximum (Ex Max) at 669 nm and an emission maximum (Em Max) at 688 nm. RR688 is designed for detection on spectral cytometers and to be excited by the Red laser (627-640 nm). It can be used on a spectral instrument in combination with APC and either R718 or Alexa Fluor 700.
TRUSTED_SUSTAINABILITY

Specifications

Applications Flow Cytometry
Classification Monoclonal
Clone B46-5
Concentration 0.2 mg/mL
Conjugate RR688
Formulation Aqueous buffered solution containing ≤0.09% sodium azide.
Gene Alias Igl@; Igl; Immunoglobulin Lambda chain
Host Species Mouse
Immunogen Pooled rat IgG mAbs
Quantity 50 μg
Regulatory Status RUO
Primary or Secondary Secondary
Target Species Rat
Content And Storage Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. The monoclonal antibody was purified from tissue culture supernatant or ascites by affinity chromatography. The antibody was conjugated to the dye under optimum conditions that minimize unconjugated dye and antibody.
Product Type Antibody
Form Liquid
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.