Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

RR688 Mouse Anti-Rat IgG2b, BD OptiBuild™

Catalog No. BDB774015 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB774015 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB774015 Supplier BD Biosciences Supplier No. 774015
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Mouse Monoclonal Secondary Antibody

Monoclonal Antibody Specifically detects in Rat Samples. It is validated for Flow Cytometry.

The G15-337 monoclonal antibody specifically recognizes rat IgG2b and does not react with other Ig isotypes. The immunogen used was a suspension of pooled rat Ig.
TRUSTED_SUSTAINABILITY

Specifications

Applications Flow Cytometry
Classification Monoclonal
Clone G15-337
Concentration 0.2 mg/mL
Conjugate RR688
Formulation Aqueous buffered solution containing ≤0.09% sodium azide.
Gene Alias Ig gamma-2B; Immunoglobulin G2b; Cγ2b
Host Species Mouse
Immunogen Pooled rat Ig
Quantity 50 μg
Regulatory Status RUO
Primary or Secondary Secondary
Target Species Rat
Content And Storage Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. The monoclonal antibody was purified from tissue culture supernatant or ascites by affinity chromatography. The antibody was conjugated to the dye under optimum conditions that minimize unconjugated dye and antibody.
Product Type Antibody
Form Liquid
Isotype IgG2b κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.