Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

RR688 Mouse Anti-Syrian Hamster IgG2, BD OptiBuild™

Catalog No. BDB772329 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB772329 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB772329 Supplier BD Biosciences Supplier No. 772329
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Mouse Monoclonal Secondary Antibody

Monoclonal Antibody Specifically detects in Syrian Hamster Samples. It is validated for Flow Cytometry.

The G192-3 clone is a Mouse BALB/c IgG1, κ isotype antibody reactive with Syrian Hamster (specifically IgG2 monoclonal antibodies, not other hamster IgG groups or IgM). It is designed for Flow Cytometry applications and is conjugated to the BD Horizon RealRedTM 688 (RR688) Dye. RR688 is a small organic fluorochrome with an excitation maximum at 669 nm and an emission maximum at 688 nm, optimized for detection on spectral cytometers excited by the Red laser (627-640 nm). The antibody is supplied at 0.2 mg/mL and is for research use only (RUO).
TRUSTED_SUSTAINABILITY

Specifications

Applications Flow Cytometry
Classification Monoclonal
Concentration 0.2 mg/mL
Conjugate RR688
Formulation Aqueous buffered solution containing ≤0.09% sodium azide.
Host Species Mouse
Immunogen Pooled Armenian and Syrian Hamster IgG mAb
Quantity 50 μg
Regulatory Status RUO
Primary or Secondary Secondary
Target Species Syrian Hamster
Content And Storage The monoclonal antibody was purified from tissue culture supernatant or ascites by affinity chromatography. The antibody was conjugated to the dye under optimum conditions that minimize unconjugated dye and antibody. Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze.
Product Type Antibody
Form Liquid
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.