Learn More
Selleck Chemical LLC RR82 Trifluoroacetate Salt
SDP
Catalog No. 5000003564
Encompass_Preferred
Pyridostatin Trifluoroacetate Salt is a G-quadruplexe stabilizer with Kd of 490 nM in a cell-free assay which targets a series of proto-oncogenes
Catalog No. 50-000-03564 Supplier Selleck Chemical LLC Supplier No. S744425MG
Pyridostatin Trifluoroacetate Salt is a G-quadruplexe stabilizer with Kd of 490 nM in a cell-free assay which targets a series of proto-oncogenes