Learn More
Aviva Systems Biology Corp RRAD Antibody - middle region (ARP56566_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP56566_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against RRAD. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human RRAD. Alternate names: RAD, RAD1, REM3ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Human, Mouse, Rabbit, Rat
Catalog No. 50-014-61908