Learn More
ABclonal Technology RRAGA Rabbit pAb
SDP
Catalog No. 5000108534
Encompass_Preferred
Enables several functions, including GTP binding activity, protein dimerization activity, and ubiquitin protein ligase binding activity. Involved in several processes, including cellular response to amino acid starvation, negative regulation of autophagy, and positive regulation of TORC1 signaling. Located in lysosome and nucleus. Colocalizes with GATOR1 complex.
Catalog No. 50-001-08534 Supplier ABclonal Technology Supplier No. A15134500UL
Enables several functions, including GTP binding activity, protein dimerization activity, and ubiquitin protein ligase binding activity. Involved in several processes, including cellular response to amino acid starvation, negative regulation of autophagy, and positive regulation of TORC1 signaling. Located in lysosome and nucleus. Colocalizes with GATOR1 complex.