Learn More
Aviva Systems Biology Corp RRAGC Antibody - N-terminal region (ARP57618_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP57618_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against RRAGC. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human RRAGC. Alternate names: FLJ13311, GTR2, RAGCApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Sheep, Zebrafish
Catalog No. 50-014-62983