Learn More
Aviva Systems Biology Corp RRAGD Antibody - middle region (ARP57542_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP57542_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against RRAGD. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human RRAGD. Alternate names: DKFZP761H171, RAGD, bA11D8.2.1ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Pig, Rabbit, Rat, Sheep, Zebrafish
Catalog No. 50-014-62855