Learn More
Aviva Systems Biology Corp RRBP1 Antibody - N-terminal region (ARP44952_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP44952_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against RRBP1. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human RRBP1. Alternate names: DKFZp586A1420, ES/130, ES130, FLJ36146, MGC157720, MGC157721, hES, RRpApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat
Catalog No. 50-014-58944