Learn More
Abnova™ RREB1/MAP3K10 Translocation FISH Probe
Catalog No. 89337350 Shop All Abnova Corporation Products
Change view
Click to view available options
For Use With (Application):
FISH-Ce
Catalog No. For Use With (Application)
89-337-350 FISH-Ce
1 options
Catalog No. 89-337-350 Supplier Abnova™ Supplier No. FT0038
Add to Cart
Add to Cart
Labeled FISH probes for identification of gene translocation using Fluoresecent In Situ Hybridization Technique.
Specifications
For Use With (Application) FISH-Ce