Learn More
CPC Scientific H-Arg-Arg-Glu-Glu-Glu-Thr-Glu-Glu-Glu-OH (trifluoroacetate salt) 1MG
SDP
Supplier:  CPC Scientific PKCP042B
Encompass
Specific substrate for casein kinase-2 (CK-2).ONE-LETTER SEQUENCE: RREEETEEEMOLECULAR FORMULA: C46H75N15O23MOLECULAR WEIGHT:1206.19STORAGE CONDITIONS: -20 5C, CAS REGISTRY NUMBER: [198481-81-1], RESEARCH AREA: Protein Kinases and PhosphatasesREFERENCES: R. Kissmehl et al., Febs Letters, 402, 227 (1997)
Catalog No. 50-211-0195