Learn More
Zymo Research Corporation RRHP 5-hmC Library Prep Kit 25 preps
SDP
Catalog No. NC1871086
Encompass_Preferred
The RRHP 5-hmC Library Prep Kit is an all-inclusive solution for analysis of genome-wide 5-hydroxymethylcytosine (5-hmC) positions at single-base resolution. The Reduced Representation Hydroxymethylation Profiling (RRHP) method is based on blocking MspI digestion by glucosylating 5-hmC within MspI recognition sites. Fragments lacking glucosylated 5-hmC at the adapter-ligation junction will be cleaved and not amplified by PCR. Therefore, only fragments containing 5-hmC will be successfully amplified and analyzed by Next-Generation Sequencing. Fragments with higher 5-hmC levels will be correlated with higher frequency of sequencing reads. RRHP bypasses the need for bisulfite conversion, which allows for DNA inputs as low as 100 ng, lower sequencing depth, and straight-forward bioinformatics processing.
Catalog No. NC1871086 Supplier Zymo Research Corporation Supplier No. D5451
The RRHP 5-hmC Library Prep Kit is an all-inclusive solution for analysis of genome-wide 5-hydroxymethylcytosine (5-hmC) positions at single-base resolution. The Reduced Representation Hydroxymethylation Profiling (RRHP) method is based on blocking MspI digestion by glucosylating 5-hmC within MspI recognition sites. Fragments lacking glucosylated 5-hmC at the adapter-ligation junction will be cleaved and not amplified by PCR. Therefore, only fragments containing 5-hmC will be successfully amplified and analyzed by Next-Generation Sequencing. Fragments with higher 5-hmC levels will be correlated with higher frequency of sequencing reads. RRHP bypasses the need for bisulfite conversion, which allows for DNA inputs as low as 100 ng, lower sequencing depth, and straight-forward bioinformatics processing.