Learn More
Aviva Systems Biology Corp RRM1 Antibody - N-terminal region (ARP46030_T100) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP46030_T100
Encompass_Preferred
This is a rabbit polyclonal antibody against RRM1. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human RRM1. Alternate names: R1, RIR1, RR1ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Yeast, Zebrafish
Catalog No. 50-014-59185