Learn More
Aviva Systems Biology Corp RRM2 Antibody - N-terminal region (ARP46031_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP46031_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against RRM2. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human RRM2. Alternate names: R2, RR2M, RR2ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Rat
Catalog No. 50-014-59186