Learn More
Cell Signaling Technology RRM2 Monoclonal Antibody, Rabbit, Human, 100 μl
SDP
Catalog No. 502732240
Encompass
The RRM2 (E7Y9J) Rabbit Monoclonal Antibody is a high-quality, recombinant antibody designed to recognize endogenous levels of total RRM2 protein. Ribonucleotide reductase subunit M2 is a critical enzyme that catalyzes the rate-limiting step in the synthesis of deoxynucleotide triphosphates. This monoclonal antibody is produced by immunizing animals with a synthetic peptide corresponding to residues surrounding Pro55 of the human RRM2 protein.
  • Validated for applications including western blotting, immunohistochemistry (paraffin), and immunofluorescence (immunocytochemistry).
  • Recognizes total endogenous RRM2 protein levels.
  • Supplied in a stable formulation containing glycerol, BSA, and sodium azide.
  • Recombinant monoclonal design ensures high specificity and consistent performance.
  • Suitable for research applications requiring detection of RRM2 in human samples.
Catalog No. 50-273-2240 Supplier Cell Signaling Technology Supplier No. 65939S
The RRM2 (E7Y9J) Rabbit Monoclonal Antibody is a high-quality, recombinant antibody designed to recognize endogenous levels of total RRM2 protein. Ribonucleotide reductase subunit M2 is a critical enzyme that catalyzes the rate-limiting step in the synthesis of deoxynucleotide triphosphates. This monoclonal antibody is produced by immunizing animals with a synthetic peptide corresponding to residues surrounding Pro55 of the human RRM2 protein.
  • Validated for applications including western blotting, immunohistochemistry (paraffin), and immunofluorescence (immunocytochemistry).
  • Recognizes total endogenous RRM2 protein levels.
  • Supplied in a stable formulation containing glycerol, BSA, and sodium azide.
  • Recombinant monoclonal design ensures high specificity and consistent performance.
  • Suitable for research applications requiring detection of RRM2 in human samples.