Learn More
Medchemexpress LLC RRM2 Human HEK293 10ug | 10ug
SDP
Catalog No. 5000197280
Encompass_Preferred
RRM2 Protein pivotal in DNA synthesis catalyzes deoxyribonucleotide biosynthesis from ribonucleotide precursors ensuring vital building blocks for DNA replication Beyond nucleotide biosynthesis RRM2 inhibits Wnt signaling indicating involvement in regulating this crucial cellular pathway RRM2 Protein Human (HEK293 His) is the recombinant human-derived RRM2 protein expressed by HEK293 with C-6 His labeled tag
Catalog No. 50-001-97280 Supplier Medchemexpress LLC Supplier No. HYP7126710UG
RRM2 Protein pivotal in DNA synthesis catalyzes deoxyribonucleotide biosynthesis from ribonucleotide precursors ensuring vital building blocks for DNA replication Beyond nucleotide biosynthesis RRM2 inhibits Wnt signaling indicating involvement in regulating this crucial cellular pathway RRM2 Protein Human (HEK293 His) is the recombinant human-derived RRM2 protein expressed by HEK293 with C-6 His labeled tag