Learn More
New England Biolabs, Inc. NEBNext rRNA Depletion Kit (Bacteria) with RNA Sample Purification Beads – 6 reactions
SDP
Catalog No. NC1775663
Encompass
The NEBNext rRNA Depletion Kit (Bacteria) employs the NEBNext RNase H-based RNA depletion workflow to target removal of rRNA from gram-positive and gram-negative organisms. The method is effective with intact and degraded RNA, from monocultures or samples with mixed bacterial species (e.g., metatranscriptomic). For convenience, this kit includes RNA Clean beads.
Catalog No. NC1775663 Supplier New England Biolabs, Inc. Supplier No. E7860S
The NEBNext rRNA Depletion Kit (Bacteria) employs the NEBNext RNase H-based RNA depletion workflow to target removal of rRNA from gram-positive and gram-negative organisms. The method is effective with intact and degraded RNA, from monocultures or samples with mixed bacterial species (e.g., metatranscriptomic). For convenience, this kit includes RNA Clean beads.