Learn More
Eurofins Discoverx ROKα/ROCK-II, active, rat, 250 µg. (14-338M)
Supplier:  Eurofins Discoverx 14338M
Encompass
N-terminal 6xHis-tagged, fusion protein corresponding to amino acids 2-543 of Rat ROKa/ROCK-II. Note: The preparation contains numerous Sf21 cellular proteins which bind weakly to Ni-NTA agarose. For detailed product information, visit www.discoverx.com/proteins.
Catalog No. 50-199-8774