Learn More
Aviva Systems Biology Corp RRP1 Antibody - C-terminal region (ARP45812_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP45812_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against RRP1. It was validated on Western Blot and immunohistochemistry. The immunogen is a synthetic peptide directed towards the C terminal region of human RRP1. Alternate names: D21S2056E, NNP-1, NOP52, RRP1AApplicationIHC, WB. Species reactivity: Human
Catalog No. 50-014-53455