Learn More
Aviva Systems Biology Corp RRP1B Antibody - middle region (ARP45850_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP45850_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against RRP1B. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human RRP1B. Alternate names: KIAA0179, Nnp1, RRP1, NNP1LApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat
Catalog No. 50-014-53456