Learn More
Aviva Systems Biology Corp RRP9 Antibody - middle region (ARP40546_T100) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP40546_T100
Encompass_Preferred
This is a rabbit polyclonal antibody against RRP9. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human RRP9. Alternate names: U3-55K, RNU3IP2ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Pig, Rabbit, Rat
Catalog No. 50-014-57517