Learn More
TARGETMOL CHEMICALS INC RRX-001 5MG
SDP
Catalog No. 502576316
Encompass_Preferred
Also available in 1 mg 2 mg 10 mg 25 mg 50 mg 100 mg 500 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. RRx-001 is a potent inhibitor ofglucose 6-phosphate dehydrogenase(G6PD) and with potent antimalarial activitypurity: 98%
Catalog No. 50-257-6316 Supplier TARGETMOL CHEMICALS INC Supplier No. T74005MG
Also available in 1 mg 2 mg 10 mg 25 mg 50 mg 100 mg 500 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. RRx-001 is a potent inhibitor ofglucose 6-phosphate dehydrogenase(G6PD) and with potent antimalarial activitypurity: 98%