Learn More
Medchemexpress LLC RS 09 | 1449566-36-2 | 99.83% | 691.78 | 10 MG
SDP
Catalog No. 5000423225
Encompass_Preferred
RS 09 is a peptide mimic of LPS and acts as a TLR4 agonist. It binds to TLR-4 and activates NF-κB, and can function as an adjuvant in vivo to enhance the antigen-specific immune response. This compound has been shown to stimulate TLR-4 and activate NF-κB in HEK-BLUE™-4 cells and induce nuclear translocation of NF-κB and secretion of inflammatory cytokines in RAW264.7 cells.
  • Acts as a TLR4 agonist
  • Binds to TLR-4 and activates NF-κB
  • Enhances antigen-specific immune response in vivo
  • Stimulates TLR-4 and activates NF-κB in HEK-BLUE™-4 cells
  • Induces nuclear translocation of NF-κB and secretion of inflammatory cytokines in RAW264.7 cells
Catalog No. 50-004-23225 Supplier Medchemexpress LLC Supplier No. HYP143910MG
RS 09 is a peptide mimic of LPS and acts as a TLR4 agonist. It binds to TLR-4 and activates NF-κB, and can function as an adjuvant in vivo to enhance the antigen-specific immune response. This compound has been shown to stimulate TLR-4 and activate NF-κB in HEK-BLUE™-4 cells and induce nuclear translocation of NF-κB and secretion of inflammatory cytokines in RAW264.7 cells.
  • Acts as a TLR4 agonist
  • Binds to TLR-4 and activates NF-κB
  • Enhances antigen-specific immune response in vivo
  • Stimulates TLR-4 and activates NF-κB in HEK-BLUE™-4 cells
  • Induces nuclear translocation of NF-κB and secretion of inflammatory cytokines in RAW264.7 cells