Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC RS 09 TFA | 99.9% | 805.80 | 10 MG
SDP

Catalog No. 5000424320
Encompass_Preferred

RS 09 TFA is an LPS peptide mimic and TLR4 agonist. It binds to TLR-4 and activates NF-κB. This compound can also function as an adjuvant in vivo, enhancing the antigen-specific immune response.

  • Stimulates TLR-4 and activates NF-κB in HEK-BLUETM-4 cells
  • Induces nuclear translocation of NF-κB and secretion of inflammatory cytokines in RAW264.7 cells
  • Enhances antigen-specific immune response in BALB/c mice
  • Molecular formula: C33H50F3N9O11
  • Appearance: Solid, white to off-white
  • Storage as powder: -80°C for 2 years, -20°C for 1 year
  • Storage in solvent: -80°C for 6 months, -20°C for 1 month (sealed, away from moisture)

Catalog No. 50-004-24320 Supplier Medchemexpress LLC Supplier No. HYP1439A10MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

RS 09 TFA is an LPS peptide mimic and TLR4 agonist. It binds to TLR-4 and activates NF-κB. This compound can also function as an adjuvant in vivo, enhancing the antigen-specific immune response.

  • Stimulates TLR-4 and activates NF-κB in HEK-BLUETM-4 cells
  • Induces nuclear translocation of NF-κB and secretion of inflammatory cytokines in RAW264.7 cells
  • Enhances antigen-specific immune response in BALB/c mice
  • Molecular formula: C33H50F3N9O11
  • Appearance: Solid, white to off-white
  • Storage as powder: -80°C for 2 years, -20°C for 1 year
  • Storage in solvent: -80°C for 6 months, -20°C for 1 month (sealed, away from moisture)

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.