Learn More
Medchemexpress LLC RS 09 TFA | 99.9% | 805.80 | 5 MG
SDP
Catalog No. 5000423916
Encompass_Preferred
RS 09 TFA is an LPS peptide mimic and TLR4 agonist that binds to TLR-4 and activates NF-κB. It functions as an adjuvant in vivo, enhancing antigen-specific immune responses.
  • Stimulates TLR-4 and activates NF-κB in HEK-BLUETM-4 cells
  • Binds to TLR-4 and induces nuclear translocation of NF-κB
  • Induces secretion of inflammatory cytokines in RAW264.7 cells
  • Increases nuclear NF-kB levels in RAW264.7 cells
  • Enhances antigen-specific immune response as an adjuvant in BALB/c mice
Catalog No. 50-004-23916 Supplier Medchemexpress LLC Supplier No. HYP1439A5MG
RS 09 TFA is an LPS peptide mimic and TLR4 agonist that binds to TLR-4 and activates NF-κB. It functions as an adjuvant in vivo, enhancing antigen-specific immune responses.
  • Stimulates TLR-4 and activates NF-κB in HEK-BLUETM-4 cells
  • Binds to TLR-4 and induces nuclear translocation of NF-κB
  • Induces secretion of inflammatory cytokines in RAW264.7 cells
  • Increases nuclear NF-kB levels in RAW264.7 cells
  • Enhances antigen-specific immune response as an adjuvant in BALB/c mice