Learn More
Selleck Chemical LLC RS-1-10mg
SDP
Supplier:  Selleck Chemical LLC S823410MG
Encompass_Preferred
RS-1 is a RAD51-stimulatory compound, which increases the DNA binding activity of RAD51. It is an HDR(homology-directed repair) enhancer that enhances Cas9- and TALEN-mediated knock-in efficiency in rabbit embryos both in vitro and in vivo.
Catalog No. 50-239-0008