Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC RS-102221 hydrochloride | 187397-18-8 | 98.0% | 649.08 | C27H32ClF3N4O7S | 10 MG
SDP

Catalog No. 5000212876
Encompass_Preferred

RS-102221 hydrochloride is the hydrochloride salt of a selective 5-HT2C receptor antagonist used in pharmacology and neuroscience research. Supplied for preclinical and in vitro studies, it provides selective blockade of 5-HT2C receptors to support receptor pharmacology, behavioral, and signaling experiments.

  • Selective 5-HT2C receptor antagonist (high affinity).
  • High purity (~98.0%).
  • Molecular weight 649.08 g/mol.
  • CAS number 187397-18-8 for unequivocal identification.
  • Available in small preclinical quantities (for example, 10 mg).
  • Product page includes datasheet and safety data sheet.

Catalog No. 50-002-12876 Supplier Medchemexpress LLC Supplier No. HY101365A10MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

RS-102221 hydrochloride is the hydrochloride salt of a selective 5-HT2C receptor antagonist used in pharmacology and neuroscience research. Supplied for preclinical and in vitro studies, it provides selective blockade of 5-HT2C receptors to support receptor pharmacology, behavioral, and signaling experiments.

  • Selective 5-HT2C receptor antagonist (high affinity).
  • High purity (~98.0%).
  • Molecular weight 649.08 g/mol.
  • CAS number 187397-18-8 for unequivocal identification.
  • Available in small preclinical quantities (for example, 10 mg).
  • Product page includes datasheet and safety data sheet.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.