Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC RS-102221 hydrochloride | 187397-18-8 | 98.0% | 649.08 g/mol | C27H32ClF3N4O7S | 1 ML
SDP

Catalog No. 5000212874
Encompass_Preferred

RS-102221 hydrochloride is a selective 5-HT2C receptor antagonist supplied as a powder and as a 10 mM solution in DMSO (1 mL). It is intended for research use in pharmacology, receptor-binding assays, and in vitro studies where a selective 5-HT2C antagonist is required.

  • Selective 5-HT2C receptor antagonist suitable for pharmacological studies.
  • High purity: 98.03% (as reported by manufacturer).
  • Available as solid (5 mg-100 mg) and as a 10 mM solution in DMSO (1 mL).
  • Molecular weight: 649.08 g/mol; chemical formula: C27H32ClF3N4O7S.
  • CAS number: 187397-18-8 for unambiguous identification.
  • Recommended storage: powder at -20°C long term; solution frozen for stability.

Catalog No. 50-002-12874 Supplier Medchemexpress LLC Supplier No. HY101365A10MM 1ML
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

RS-102221 hydrochloride is a selective 5-HT2C receptor antagonist supplied as a powder and as a 10 mM solution in DMSO (1 mL). It is intended for research use in pharmacology, receptor-binding assays, and in vitro studies where a selective 5-HT2C antagonist is required.

  • Selective 5-HT2C receptor antagonist suitable for pharmacological studies.
  • High purity: 98.03% (as reported by manufacturer).
  • Available as solid (5 mg-100 mg) and as a 10 mM solution in DMSO (1 mL).
  • Molecular weight: 649.08 g/mol; chemical formula: C27H32ClF3N4O7S.
  • CAS number: 187397-18-8 for unambiguous identification.
  • Recommended storage: powder at -20°C long term; solution frozen for stability.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.