Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Rs-102221 hydrochloride | 187397-18-8 | 98.0% | 649.08 g/mol | C27H32ClF3N4O7S | 100 MG
SDP

Catalog No. 5000211810
Encompass_Preferred

RS-102221 hydrochloride is a selective 5-HT2C receptor antagonist provided as a solid research reagent for neuroscience and pharmacology studies. It is utilized to investigate 5-HT2C receptor function in vitro and in vivo and has been reported to promote neuronal differentiation and increase food intake and weight gain in animal models.

  • Selective 5-HT2C receptor antagonist for research applications.
  • Purity 98.03% as supplied.
  • Molecular weight 649.08 g/mol; formula C27H32ClF3N4O7S.
  • Appearance: solid, white to off-white.
  • Storage: powder -20°C (3 years) or 4°C (2 years); in solvent -80°C (6 months).
  • Available in multiple sizes, including 100 mg.

Catalog No. 50-002-11810 Supplier Medchemexpress LLC Supplier No. HY101365A100MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

RS-102221 hydrochloride is a selective 5-HT2C receptor antagonist provided as a solid research reagent for neuroscience and pharmacology studies. It is utilized to investigate 5-HT2C receptor function in vitro and in vivo and has been reported to promote neuronal differentiation and increase food intake and weight gain in animal models.

  • Selective 5-HT2C receptor antagonist for research applications.
  • Purity 98.03% as supplied.
  • Molecular weight 649.08 g/mol; formula C27H32ClF3N4O7S.
  • Appearance: solid, white to off-white.
  • Storage: powder -20°C (3 years) or 4°C (2 years); in solvent -80°C (6 months).
  • Available in multiple sizes, including 100 mg.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.