Learn More
Medchemexpress LLC RS-102221 hydrochloride | 187397-18-8 | 98.0% | 649.08 g/mol | C27H32ClF3N4O7S | 25 MG
SDP
Catalog No. 5000212438
Encompass_Preferred
RS-102221 hydrochloride is the hydrochloride salt of a selective 5-HT2C receptor antagonist commonly used in neuroscience research. It demonstrates high affinity for 5-HT2C receptors and is supplied in both solid and solution forms for in vitro and in vivo studies, with specified purity and storage recommendations.
Catalog No. 50-002-12438 Supplier Medchemexpress LLC Supplier No. HY101365A25MG
RS-102221 hydrochloride is the hydrochloride salt of a selective 5-HT2C receptor antagonist commonly used in neuroscience research. It demonstrates high affinity for 5-HT2C receptors and is supplied in both solid and solution forms for in vitro and in vivo studies, with specified purity and storage recommendations.