Learn More
Medchemexpress LLC RS-102221 hydrochloride | 187397-18-8 | 98.0% | 649.08 g/mol | C27H32ClF3N4O7S | 5 MG

Supplier: Medchemexpress LLC HY101365A5MG
RS-102221 hydrochloride is the hydrochloride salt of a selective 5-HT2C receptor antagonist supplied as a solid for research use in pharmacology and neuroscience studies. It is intended for receptor binding and functional assays and includes manufacturer guidance for storage and solubility to support reliable preparation.
- Selective 5-HT2C receptor antagonist for pharmacology research.
- White to off-white solid suitable for in vitro and in vivo studies.
- High purity: 98.0%.
- Molecular weight: 649.08 g/mol.
- Soluble in DMSO at 125 mg/mL (ultrasonic recommended).
- Store powder at -20°C for long term; stock solutions at -80°C.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.